APO455838090 Apollo Scientific Ltd

2-Amino-4-(trifluoromethyl)-1,3-thiazole

  • MDL number MFCD00832759
  • Cas Number : 349-49-5
  • Molecular Weight : 168.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-70
Synonym 4-(Trifluoromethyl)-1,3-thiazol-2-amine
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethyl-13-thiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethyl-13-thiazole
pictograms GHS07
hstatements H315;H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart