APO455838093 Apollo Scientific Ltd

4-Bromo-1,1,1,3,3-pentafluorobutane

  • MDL number MFCD08460470
  • Cas Number : 933600-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,4,4,4-Pentafluorobutyl bromide
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38433

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38433
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart