APO455838095 Apollo Scientific Ltd

4-(Trifluoromethyl)benzophenone

  • MDL number MFCD00000400
  • Cas Number : 728-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-119
Synonym Phenyl[4-(trifluoromethyl)phenyl]methanone, 4-Benzoylbenzotrifluoride
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5287

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5287
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart