APO455838100 Apollo Scientific Ltd

2,6-Difluoropyridine-3-boronic acid

  • MDL number MFCD03788245
  • Cas Number : 136466-94-9
  • Molecular Weight : 158.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-98
Synonym 3-Borono-2,6-difluoropyridine
Purity 96
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoropyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoropyridine-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart