APO455838104 Apollo Scientific Ltd

3-Bromo-4-fluorobenzamide

  • MDL number MFCD03094431
  • Cas Number : 455-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-fluorobenzamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-fluorobenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart