APO455838108 Apollo Scientific Ltd

4-Fluoro-2-hydroxyphenacyl bromide

  • MDL number MFCD09763689
  • Cas Number : 866863-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37212

Safety Information

signalword Danger
pcodes P280;P301+P330+P331
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37212
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart