APO455838110 Apollo Scientific Ltd

1-[2,6-Bis(trifluoromethyl)quinol-4-yl]piperazine

  • MDL number MFCD03094433
  • Cas Number : 401567-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2398

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2398
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart