APO455838111 Apollo Scientific Ltd

2,6-Difluoro-N-methoxy-N-methylbenzamide

  • MDL number MFCD08689758
  • Cas Number : 937601-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-54
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-n-methoxy-n-methylbenzamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-n-methoxy-n-methylbenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart