APO455838112 Apollo Scientific Ltd

Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

  • MDL number MFCD06656414
  • Cas Number : 128073-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-2-(ethoxycarbonyl)-5-(trifluoromethyl)pyridine, Ethyl 3-chloro-5-(trifluoromethyl)picolinate
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37315

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=37315
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart