APO455838126 Apollo Scientific Ltd

5-(4-Fluorophenyl)-3,4-dihydro-2H-1,4-thiazine-3-carboxylic acid

  • MDL number MFCD08689744
  • Cas Number : 1190102-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-132
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37319

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37319
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart