APO455838128 Apollo Scientific Ltd

N-Methoxy-N-methyl-3-(trifluoromethyl)benzamide

  • MDL number MFCD08689665
  • Cas Number : 116332-62-8
  • Molecular Weight : 233.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-[Methoxy(methyl)carbamoyl]benzotrifluoride
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-methoxy-n-methyl-3-trifluoromethylbenzamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-methoxy-n-methyl-3-trifluoromethylbenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart