APO455838129 Apollo Scientific Ltd

N-Methoxy-N-methyl-4-(trifluoromethyl)benzamide

  • MDL number MFCD08689795
  • Cas Number : 116332-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[Methoxy(methyl)carbamoyl]benzotrifluoride, 4-{[Methoxy(methyl)amino]carbonyl}benzotrifluoride
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37323

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=37323
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart