APO455838133 Apollo Scientific Ltd

Methyl 3-amino-4-fluorobenzoate

  • MDL number MFCD08689743
  • Cas Number : 369-26-6
  • Molecular Weight : 169.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-69
Synonym 2-Fluoro-5-(methoxycarbonyl)aniline
Purity 99
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-amino-4-fluorobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-amino-4-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart