APO455838134 Apollo Scientific Ltd

6-Bromo-3-chloro-2,4-difluoroaniline

  • MDL number MFCD00143495
  • Cas Number : 201849-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2405

Safety Information

signalword Warning
pcodes P280;P271;P260
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2405
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart