APO455838136 Apollo Scientific Ltd

3-(Trifluoromethyl)butanoic acid

  • MDL number MFCD00039533
  • Cas Number : 348-75-4
  • Molecular Weight : 156.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Trifluoromethyl)butyric acid, 3-Methyl-4,4,4-trifluorobutanoic acid
Purity 96
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 91/25mm
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethylbutanoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethylbutanoic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart