APO455838149 Apollo Scientific Ltd

3,5-Difluoro-2-nitroanisole

  • MDL number MFCD11865203
  • Cas Number : 66684-61-5
  • Molecular Weight : 189.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 59.5-59.9
Iupac Name 1,5-difluoro-3-methoxy-2-nitrobenzene
Inchi Code 1S/C7H5F2NO3/c1-13-6-3-4(8)2-5(9)7(6)10(11)12/h2-3H,1H3
Inchi Key DXEGOHULGVHKCT-UHFFFAOYSA-N

Description

Synonym 1,5-Difluoro-3-methoxy-2-nitrobenzene, 3,5-Difluoro-2-nitrophenyl methyl ether
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 59.5-59.9
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-2-nitroanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-2-nitroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart