APO455838150 Apollo Scientific Ltd

2',4'-Difluoro-3'-(trifluoromethyl)acetophenone

  • MDL number MFCD12547946
  • Cas Number : 1202679-46-6
  • Molecular Weight : 224.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[2,4-Difluoro-3-(trifluoromethyl)phenyl]ethan-1-one, 3-Acetyl-2,6-difluorobenzotrifluoride
Purity 98
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-difluoro-3-trifluoromethylacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-difluoro-3-trifluoromethylacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart