APO455838151 Apollo Scientific Ltd

2-(Trifluoromethyl)-DL-lactic acid

  • MDL number MFCD00190646
  • Cas Number : 374-35-6
  • Molecular Weight : 158.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 85
Iupac Name 3,3,3-trifluoro-2-hydroxy-2-methylpropanoic acid
Inchi Code 1S/C4H5F3O3/c1-3(10,2(8)9)4(5,6)7/h10H,1H3,(H,8,9)
Inchi Key CTGJACFEVDCYMC-UHFFFAOYSA-N

Description

Synonym 2-Hydroxy-2-methyl-3,3,3-trifluoropropionic acid, 2-Hydroxy-2-(trifluoromethyl)propanoic acid
Purity 97%
Flash Point 189
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 85
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethyl-dl-lactic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethyl-dl-lactic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart