APO455838153 Apollo Scientific Ltd

2-(Trifluoromethyl)maleic anhydride

  • MDL number MFCD00153287
  • Cas Number : 700-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Trifluoromethyl)furan-2,5-dione, 2,5-Dihydro-2,5-dioxo-3-(trifluoromethyl)furan
Purity 97
Flash Point 37
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 93/65mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5346

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5346
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart