APO455838164 Apollo Scientific Ltd

5-Fluoro-2-methoxypyridine-3-boronic acid

  • MDL number MFCD07368239
  • Cas Number : 957120-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-88
Synonym 3-Borono-5-fluoro-2-methoxypyridine
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-methoxypyridine-3-boronic-acid-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-methoxypyridine-3-boronic-acid-98
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart