APO455838171 Apollo Scientific Ltd

3-Bromo-4-(trifluoromethyl)pyridine

  • MDL number MFCD08061310
  • Cas Number : 936841-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-alpha,alpha,alpha-trifluoro-4-picoline
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 58/11mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart