APO455838182 Apollo Scientific Ltd

5-Fluoro-2-methoxybenzyl alcohol

  • MDL number MFCD00671767
  • Cas Number : 426831-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37416

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=37416
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart