APO455838184 Apollo Scientific Ltd

2-Bromo-5-fluorobenzoyl chloride

  • MDL number MFCD03094438
  • Cas Number : 111771-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20-22
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2887

Safety Information

signalword Danger
pcodes P280;P260;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2887
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart