APO455838185 Apollo Scientific Ltd

2-Bromo-5-fluorophenylacetonitrile

  • MDL number MFCD03094439
  • Cas Number : 886761-96-2
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-71
Synonym 2-Bromo-5-fluorobenzyl cyanide, (2-Bromo-5-fluorophenyl)acetonitrile
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-fluorophenylacetonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-fluorophenylacetonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart