APO455838188 Apollo Scientific Ltd

2-Chloro-5-fluorobromobenzene

  • MDL number MFCD00061120
  • Cas Number : 201849-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-chloro-4-fluorobenzene
Purity 99
Density 1.75
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-fluorobromobenzene-99

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-fluorobromobenzene-99
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart