APO455838191 Apollo Scientific Ltd

4-Amino-7-(trifluoromethyl)quinoline

  • MDL number MFCD00269411
  • Cas Number : 243666-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=161

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=161
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart