APO455838194 Apollo Scientific Ltd

2-Methyl-3,3,3-trifluoropropanal

  • MDL number MFCD00042425
  • Cas Number : 58928-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Trifluoromethyl)propanal, 2-(Trifluoromethyl)propionaldehyde, 2-Methyl-3,3,3-trifluoropropaldehyde
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 66
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8179

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8179
pictograms GHS02;GHS07
hstatements H225;H302;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart