APO455838195 Apollo Scientific Ltd

2-Amino-5-(trifluoromethoxy)benzaldehyde, N-BOC protected

  • MDL number MFCD08669635
  • Cas Number : 505084-60-6
  • Molecular Weight : 305.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-104
Synonym tert-Butyl [2-formyl-4-(trifluoromethoxy)phenyl]carbamate, 2-[(tert-Butoxycarbonyl)amino]-5-(trifluoromethoxy)benzaldehyde
Purity 94
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-trifluoromethoxybenzaldehyde-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-trifluoromethoxybenzaldehyde-n-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart