APO455838200 Apollo Scientific Ltd

3-(Trifluoromethyl)-2-pyrazolin-5-one

  • MDL number MFCD00052748
  • Cas Number : 401-73-0
  • Molecular Weight : 152.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-217
Purity 97
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-trifluoromethyl-2-pyrazolin-5-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-trifluoromethyl-2-pyrazolin-5-one
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart