APO455838206 Apollo Scientific Ltd

4-(Trifluoromethyl)thiobenzamide

  • MDL number MFCD00051806
  • Cas Number : 72505-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-138
Purity 98
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5517

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5517
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart