APO455838207 Apollo Scientific Ltd

2,4-Difluorobenzotrifluoride

  • MDL number MFCD04972808
  • Cas Number : 64248-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4-Difluoro-1-(trifluoromethyl)benzene, alpha,alpha,alpha,2,4-Pentafluorotoluene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47126

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47126
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart