APO455838210 Apollo Scientific Ltd

4-(Trifluoromethyl)thiophenol

  • MDL number MFCD00042320
  • Cas Number : 825-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzenethiol, 4-(Trifluoromethyl)phenyl mercaptan
Purity 97
Density 1.3
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 66-68/15mm
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethylthiophenol-97

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethylthiophenol-97
pictograms GHS07
hstatements H312;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart