APO455838212 Apollo Scientific Ltd

Ethyl 2-chloro-4-fluorobenzoate

  • MDL number MFCD06204388
  • Cas Number : 167758-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 239-241
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8798

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=8798
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart