APO455838214 Apollo Scientific Ltd

Ethyl 4-chloro-2-fluorobenzoate

  • MDL number MFCD03095401
  • Cas Number : 4793-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 206
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8800

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8800
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart