APO455838218 Apollo Scientific Ltd

3,5-Dichloro-2,4-difluoroaniline

  • MDL number MFCD00044104
  • Cas Number : 83121-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75
Synonym 1-Amino-3,5-dichloro-2,4-difluorobenzene
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/35-dichloro-24-difluoroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloro-24-difluoroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart