APO455838225 Apollo Scientific Ltd

Ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propanoate

  • MDL number MFCD00523020
  • Cas Number : 94695-50-8
  • Molecular Weight : 264.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48
Synonym Ethyl 3-oxo-3-(2,3,4,5-tetrafluorophenyl)propionate, Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 100/0.075mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-oxo-3-2345-tetrafluorophenylpropanoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-oxo-3-2345-tetrafluorophenylpropanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart