APO455838229 Apollo Scientific Ltd

5-Chloro-2-fluoronitrobenzene

  • MDL number MFCD00069418
  • Cas Number : 345-18-6
  • Molecular Weight : 175.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 8
Synonym 4-Chloro-1-fluoro-2-nitrobenzene
Purity 97
Flash Point 113
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 237
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-fluoronitrobenzene
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart