APO455838233 Apollo Scientific Ltd

5,6-Difluoro-1H-indole

  • MDL number MFCD01075212
  • Cas Number : 169674-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6066

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=6066
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart