APO455838237 Apollo Scientific Ltd

1-(3,4-Difluorophenyl)biguanide hydrochloride

  • MDL number MFCD03094449
  • Cas Number : 537037-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-215
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6071

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=6071
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart