APO455838247 Apollo Scientific Ltd

3,4,5-Trifluorophenylacetonitrile

  • MDL number MFCD00083531
  • Cas Number : 220228-03-5
  • Molecular Weight : 171.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28-34
Synonym 3,4,5-Trifluorobenzyl cyanide
Purity 97
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/345-trifluorophenylacetonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/345-trifluorophenylacetonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart