APO455838253 Apollo Scientific Ltd

Dichloro(methyl)(3,3,3-trifluoroprop-1-yl)silane

  • MDL number MFCD00039294
  • Cas Number : 675-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.261
Flash Point 15
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 121-122
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5611

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=5611
pictograms GHS02;GHS05;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart