APO455838254 Apollo Scientific Ltd

1-[4-(Trifluoromethoxy)phenyl]biguanide hydrochloride

  • MDL number MFCD01569536
  • Cas Number : 42823-09-6
  • Molecular Weight : 297.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-213
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-trifluoromethoxyphenylbiguanide-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-trifluoromethoxyphenylbiguanide-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart