APO455838258 Apollo Scientific Ltd

2,3-Dimethyl-4-fluoronitrobenzene

  • MDL number MFCD05663712
  • Cas Number : 1736-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37664

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=37664
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart