APO455838259 Apollo Scientific Ltd

Methyl 2-nitro-4-(trifluoromethyl)benzoate

  • MDL number MFCD08282772
  • Cas Number : 228418-45-9
  • Molecular Weight : 249.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-nitro-4-(trifluoromethyl)benzoate
Inchi Code 1S/C9H6F3NO4/c1-17-8(14)6-3-2-5(9(10,11)12)4-7(6)13(15)16/h2-4H,1H3
Inchi Key WYTRZKAYPKZGCN-UHFFFAOYSA-N

Description

Synonym 4-(Methoxycarbonyl)-3-nitrobenzotrifluoride, Methyl 2-nitro-alpha,alpha,alpha-trifluoro-p-toluate
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-nitro-4-trifluoromethylbenzoate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/methyl-2-nitro-4-trifluoromethylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart