APO455838263 Apollo Scientific Ltd

5-Chloro-2-fluorobenzotrifluoride

  • MDL number MFCD00060655
  • Cas Number : 89634-74-2
  • Molecular Weight : 198.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-1-fluoro-2-(trifluoromethyl)benzene, 5-Chloro-alpha,alpha,alpha,2-tetrafluorotoluene
Purity 98
Density 1.43
Flash Point 47
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 145
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-fluorobenzotrifluoride
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart