APO455838266 Apollo Scientific Ltd

2-Fluoro-5-nitrophenylacetic acid

  • MDL number MFCD04091773
  • Cas Number : 195609-18-8
  • Molecular Weight : 199.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Synonym 3-(Carboxymethyl)-4-fluoronitrobenzene
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-nitrophenylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-nitrophenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart