APO455838267 Apollo Scientific Ltd

2-Fluoro-4-methylbenzotrifluoride

  • MDL number MFCD07368763
  • Cas Number : 158364-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.24
Flash Point 39
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 146
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9460

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=9460
pictograms GHS02;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart