APO455838268 Apollo Scientific Ltd

4,5,6-Trifluoropyrimidine

  • MDL number MFCD05662363
  • Cas Number : 17573-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,5,6-Trifluoro-1,3-diazine
Density 1.4609
Flash Point 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 83-85
Product Link https://store.apolloscientific.co.uk/product/456-trifluoropyrimidine

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/456-trifluoropyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart