APO455838271 Apollo Scientific Ltd

4-Hydroxy-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD01091008
  • Cas Number : 220239-68-9
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-147
Synonym 4-Carboxy-2-(trifluoromethyl)phenol, 5-Carboxy-2-hydroxybenzotrifluoride
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-3-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-3-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart