APO455838273 Apollo Scientific Ltd

4,4,4-Trifluoro-3-(trifluoromethyl)butanoic acid

  • MDL number MFCD00236630
  • Cas Number : 17327-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,3-Bis(trifluoromethyl)propanoic acid, 4,4,4-Trifluoro-3-(trifluoromethyl)butyric acid
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8181

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8181
pictograms GHS05;GHS07
hstatements H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart